Merck
  • Evaluation of broad-spectrum anthelmintic activity in a novel assay against Haemonchus contortus, Trichostrongylus colubriformis and T. sigmodontis in the gerbil Meriones unguiculatus.
Evaluation of broad-spectrum anthelmintic activity in a novel assay against Haemonchus contortus, Trichostrongylus colubriformis and T. sigmodontis in the gerbil Meriones unguiculatus.
Journal of helminthology (2006-11-28)
D A Ostlind, S Cifelli, W G Mickle, S K Smith, D V Ewanciw, B Rafalko, T Felcetto, A Misura
ZUSAMMENFASSUNG
The gerbil Meriones unguiculatus, infected with three species of nematodes, each located in a separate part of the gastrointestinal tract, provided a reliable laboratory assay for the evaluation of broad-spectrum anthelmintic activity. Gerbils harbouring 6-day-old infections of Haemonchus contortus, Trichostrongylus colubriformis and T. sigmodontis were given selected broad-spectrum anthelmintics by gavage. Three benzimidazoles, thiabendazole, oxfendazole and albendazole, a tetrahydropyrimidine, morantel, an imidazothiazole, levamisole hydrochloride, a macrocyclic lactone, ivermectin and an experimental natural product, paraherquamide, were active against all three nematodes at various dosages. Trichostrongylus colubriformis was most sensitive to levamisole hydrochloride, morantel, thiabendazole and paraherquamide whereas ivermectin, oxfendazole and albendazole were more effective against H. contortus. All compounds were active against the caecal nematode T. sigmodontis although it was less sensitive than T. colubriformis. Haemonchus contortus was more sensitive than T. sigmodontis to all anthelmintics tested except thiabendazole.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Oxfendazol, VETRANAL®, analytical standard
Supelco
Morantel-tartrat Hydrat, VETRANAL®, analytical standard