Merck
  • Application of extraction chromatography to the separation of thorium and uranium dissolved in a solution of high salt concentration.
Application of extraction chromatography to the separation of thorium and uranium dissolved in a solution of high salt concentration.
Journal of chromatography. A (2006-12-13)
Asako Fujiwara, Yutaka Kameo, Akiko Hoshi, Tomoko Haraga, Mikio Nakashima
ZUSAMMENFASSUNG
Extraction chromatography with commercially available UTEVA resin (for uranium and tetravalent actinide) was applied for the separation of Th and U from control solutions prepared from a multi-element control solution and from sample solutions of solidified simulated waste. Thorium and U in control solutions with 1-5mol/dm(3) HNO(3) were extracted with UTEVA resin and recovered with a solution containing 0.1mol/dm(3) HNO(3) and 0.05mol/dm(3) oxalic acid to be separated from the other metallic elements. Extraction behavior of U in the sample solutions was similar to that in the control solutions, but extraction of Th was dependent on the concentration of HNO(3). Thorium was extracted from 5mol/dm(3) HNO(3) sample solutions but not from 1mol/dm(3) HNO(3) sample solutions. We conjecture that thorium fluoride formation interferes with extraction of Th. Addition of Al(NO(3))(3) and Fe(NO(3))(3), which have higher stability constant with fluoride ion than Th, does improve extractability of Th from 1mol/dm(3) HNO(3) sample solution.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumnitrat Nonahydrat, ACS reagent, ≥98%
Sigma-Aldrich
Aluminiumnitrat Nonahydrat, 99.997% trace metals basis