Merck
  • Conformational dynamics of substrate in the active site of cytochrome P450 BM-3/NPG complex: insights from NMR order parameters.
Conformational dynamics of substrate in the active site of cytochrome P450 BM-3/NPG complex: insights from NMR order parameters.
Journal of the American Chemical Society (2007-01-18)
Krishna Pratap Ravindranathan, Emilio Gallicchio, Ann E McDermott, Ronald M Levy
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PalGly, ≥98% (HPLC), solid