Merck
Behavioral syndrome induced by allylnitrile, crotononitrile or 2-pentenenitrile in rats.
Neuropharmacology (1991-08-01)
H Tanii, M Hayashi, K Hashimoto
ZUSAMMENFASSUNG
A single oral administration of allylnitrile, crotononitrile or 2-pentenenitrile in rats induced behavioral abnormalities, such as head-twitching, head weaving, hindlimb abduction, backward pedaling and pivoting. The head-twitching, which was most consistently observed, was suppressed by serotonin (5-HT) antagonists, cyproheptadine or methysergide or by the 5-HT depleter, dl-p-chlorophenylalanine but was accentuated by the 5-HT releaser, dl-p-chloroamphetamine. The results suggest that the 5-HT system is involved in producing the behavioral abnormalities. To discover the effects of allylnitrile, crotononitrile and 2-pentenenitrile on the metabolism of 5-HT and dopamine, 6 areas of the brain of the rat were examined on days 1, 6, 15 and 30 after injection. Each of the nitriles caused significant increases in the level of 5-HT and 5-hydroxyindoleacetic acid (5-HIAA) and in the ratio of 5-HIAA/5-HT, one day after injection. The increase in 5-HIAA was most remarkable, suggesting an enhancement of the serotonergic system. The three nitriles had no effect on the metabolism of dopamine, over a period of 30 days.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Crotonsäurenitril (cis+trans), 99%