Merck
  • Calcium influx through L-type channels generates protein kinase M to induce burst firing of dopamine cells in the rat ventral tegmental area.
Calcium influx through L-type channels generates protein kinase M to induce burst firing of dopamine cells in the rat ventral tegmental area.
The Journal of biological chemistry (2007-01-24)
Yudan Liu, Jules Dore, Xihua Chen
ZUSAMMENFASSUNG
Enhanced activity of the dopaminergic system originating in the ventral tegmental area is implicated in addictive and psychiatric disorders. Burst firing increases dopamine levels at the synapse to signal novelty and salience. We have previously reported a calcium-dependent burst firing of dopamine cells mediated by L-type channels following cholinergic stimulation; this paper describes a cellular mechanism resulting in burst firing following L-type channel activation. Calcium influx through L-type channels following FPL 64176 or (S)-(-)-Bay K8644 induced burst firing independent of dopamine, glutamate, or calcium from the internal stores. Burst firing induced as such was completely blocked by the substrate site protein kinase C (PKC) inhibitor chelerythrine but not by the diacylglycerol site inhibitor calphostin C. Western blotting analysis showed that FPL 64176 and (S)-(-)-Bay K8644 increased the cleavage of PKC to generate protein kinase M (PKM) and the specific calpain inhibitor MDL28170 blocked this increase. Prevention of PKM production by inhibiting calpain or depleting PKC blocked burst firing induction whereas direct loading of purified PKM into cells induced burst firing. Activation of the N-methyl-D-aspartic acid type glutamate or cholinergic receptors known to induce burst firing increased PKM expression. These results indicate that calcium influx through L-type channels activates a calcium-dependent protease that cleaves PKC to generate constitutively active and labile PKM resulting in burst firing of dopamine cells, a pathway that is involved in glutamatergic or cholinergic modulation of the central dopamine system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
FPL 64176, ≥98% (HPLC), powder