Merck
  • Visible-light induced hydrogen production using a polypeptide-chlorophyll complex with alpha-helix conformation.
Visible-light induced hydrogen production using a polypeptide-chlorophyll complex with alpha-helix conformation.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2007-02-06)
Pascaline Ngweniform, Yoshihumi Kusumoto, Takeo Teshima, Miyuki Ikeda, Shouichi Somekawa, Bashir Ahmmad
ZUSAMMENFASSUNG
Hydrogen production was accomplished under visible-light irradiation by using a system consisting of a biomolecule (chlorophyll a), methylviologen, ethylenediaminetetraacetic acid disodium salt and Pt-loaded poly(l-glutamate) (Poly(Glu)), in aqueous decylammonium chloride (DeAC) solution. Spectroscopic studies revealed that chlorophyll a is solubilized in the hydrophobic clusters of Pt-loaded Poly(Glu)-decylammonium chloride. In the Poly(Glu)-DeAC complex, the electron transfer occurred between chlorophyll a and methylviologen leading to hydrogen production. The most noticeable result is that the rate of hydrogen evolution depends on the change from the random coil to the alpha-helix in conformation of Poly(Glu) induced by the cooperative binding with DeAC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Decylamin, 95%
Sigma-Aldrich
Decylamin, ≥99.0% (GC)