Merck
Spirodiepoxide reaction with cuprates.
Journal of the American Chemical Society (2007-02-10)
Partha Ghosh, Stephen D Lotesta, Lawrence J Williams
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumbromid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithiumbromid -Lösung, 54 wt. % in H2O
Sigma-Aldrich
Lithiumbromid, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
Lithiumbromid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithiumbromid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis