Merck
  • Effects of water and diet acidification with and without antibiotics on weanling pig growth and microbial shedding.
Effects of water and diet acidification with and without antibiotics on weanling pig growth and microbial shedding.
Journal of animal science (2007-02-14)
M C Walsh, D M Sholly, R B Hinson, K L Saddoris, A L Sutton, J S Radcliffe, R Odgaard, J Murphy, B T Richert
ZUSAMMENFASSUNG
Two 5-wk experiments were conducted to determine the effects of water and diet acidification with and without antibiotics on weanling pig growth performance and microbial shedding. In Exp. 1, 204 pigs (19.2 d of age) were used in a 3 x 2 factorial, with 3 dietary treatments fed with or without water acidification (2.58 mL/L of a propionic acid blend; KEM SAN, Kemin Americas, Des Moines, IA). Dietary treatments were: 1) control, 2) control + 55 ppm of carbadox (CB), and 3) dietary acid [DA; control + 0.4% organic acid-based blend (fumaric, lactate, citric, propionic, and benzoic acids; Kemin Americas)] on d 0 to 7 followed by 0.2% inorganic acid-based blend (phosphoric, fumaric, lactic, and citric acids; Kemin Americas) on d 7 to 34. In Exp. 2, 210 pigs (average 18.3 d of age) were fed 1 of 3 dietary treatments: 1) control, 2) control + 55 ppm of CB, and 3) control + 38.6 ppm of tiamulin + 441 ppm of chlortetracycline on d 0 to 7 followed by 110 ppm of chlortetracycline on d 7 to 35 (TC) with or without dietary acidification (same as Exp. 1) in a 3 x 2 factorial arrangement of treatments. For both experiments, the pigs were allotted based on genetics, sex, and initial BW [5.5 kg (Exp. 1) or 5.6 kg (Exp. 2)]. Pigs were housed at 6 or 7 (Exp. 1) and 7 (Exp. 2) pigs/pen. Treatments were fed in 3 phases: d 0 to 7, 7 to 21, and 21 to 35 (34 d, Exp. 1). Fecal grab samples were collected from 3 pigs/pen on d 6, 20, and 33 for measurement of pH and Escherichia coli. During phase 3 and overall in Exp. 1, pigs fed CB had greater (P < 0.001) ADG (overall ADG, 389 vs. 348, and 348 g/d, respectively), ADFI (P < 0.007, 608 vs. 559, and 554 g/d, respectively), and d 34 BW (P < 0.001, 18.8 vs. 17.3, and 17.3 kg, respectively) than pigs fed NC and DA. Phase 3 ADG was improved (P < 0.01) by water acidification across all diets. In Exp. 2, pigs fed CB and TC had greater ADG (P < 0.004; 315 and 303 vs. 270 g/d, respectively), ADFI (P < 0.01), and d 35 BW (P < 0.002; 16.7 and 16.2 vs. 15.1 kg, respectively) than pigs fed NC. There was a tendency (P < 0.08) for an improvement in ADG when DA was added to the NC or TC, but decreased ADG when DA was added to CB.