Merck
  • Five oximes (K-27, K-48, obidoxime, HI-6 and trimedoxime) in comparison with pralidoxime: survival in rats exposed to methyl-paraoxon.
Five oximes (K-27, K-48, obidoxime, HI-6 and trimedoxime) in comparison with pralidoxime: survival in rats exposed to methyl-paraoxon.
Journal of applied toxicology : JAT (2007-02-17)
G A Petroianu, S M Nurulain, N Nagelkerke, M Shafiullah, J Kassa, K Kuca
ZUSAMMENFASSUNG
There is a clear need for broad-spectrum cholinesterase reactivators (active against a multitude of organophosphorus ester enzyme inhibitors) with a higher efficacy than pralidoxime. The purpose of the study was to quantify in vivo the extent of oxime-conferred protection, using methyl-paraoxon [dimethyl p-nitrophenyl phosphate; (methyl-POX)] as a cholinesterase inhibitor. There were seven groups of six rats in each cycle of the experiment. Group 1 (G1) received 2 micromol methyl-POX ( approximately LD(50)), the other groups (G2-7) received 2 micromol methyl-POX + one of the six reactivators. The animals were monitored for 48 h and the time of mortality was recorded. The procedure was repeated six times. All substances were applied i.p. The experiments were repeated using 3 and 5 micromol methyl-POX. Mortality data were compared and hazards ratios (relative risks) ranked using the Cox proportional hazards model with methyl-POX dose and group (reactivator) as time-independent covariables. The relative risk of death estimated by Cox analysis (95% CI) in oxime-treated animals when compared with untreated animals, adjusted for methyl-POX dose (high/low) was K-27, 0.58 (0.42-0.80); K-48, 0.60 (0.43-0.83); trimedoxime, 0.76 (0.55-1.04); pralidoxime, 0.88 (0.65-1.20); obidoxime, 0.93 (0.68-1.26); HI-6, 0.96 (0.71-1.31). Only K-27 and K-48 provided statistically significant protection in rats exposed to methyl-POX. Despite the lower inhibitory potency (higher IC(50)) of methyl-POX compared with POX (ratio 4:1), the ability of oxime reactivators to protect from methyl-POX induced mortality was reduced compared with protection from POX (ethyl-analog).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Paraoxonmethyl, PESTANAL®, analytical standard