Merck
The skin sensitization potential of resorcinol: experience with the local lymph node assay.
Contact dermatitis (2007-03-09)
David A Basketter, David Sanders, Ian R Jowsey
ZUSAMMENFASSUNG
Resorcinol is a simple aromatic chemical (1,3-benzenediol) that has found widespread use, particularly as a coupler in hair dyes. Clinical experience clearly shows that resorcinol is a (albeit uncommon) skin sensitizer. By contrast, predictive methods, both animal and human, have previously failed to identify resorcinol as such. Here, we describe the outcome of a recent local lymph node assay performed in accordance with Organisation for Economic Co-operation and Development guideline 429, which correctly identified resorcinol as a skin sensitizer. Clear evidence of a dose response was apparent, and an EC3 value of approximately 6% was calculated. This suggests that the skin-sensitizing potency of resorcinol is approximately 2 orders of magnitude lower than that of p-phenylenediamine but similar to that of hexyl cinnamic aldehyde. These data show the importance of adherence to test guidelines and aligns the clinical experience with resorcinol with that obtained in predictive animal methods.