Merck
Decomposition of phorate in aqueous solution by ozonation.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2007-03-17)
Young Ku, Hung Shian Lin, Wen Wang, Chih Ming Ma
ZUSAMMENFASSUNG
Phorate (O,O-diethyl S-ethylthiomethyl phosphorodithioate) dissolved in aqueous solution was almost completely decomposed by ozonation to form various species within 10 minutes of reaction time for the experimental conditions examined in this research. The generation rate of sulfate was found to be fairly independent of solution pH value. However, the formation of phosphate and carbonate was more favorable for alkaline solutions where hydroxyl free radical is the primary oxidative species. The reaction rates increased with initial gaseous ozone concentrations, indicating the reaction was mass transfer-controlled within the experimental range of this research. Combining the analytical results by various instruments, including gas chromatograph equipped with an electron ionization detector (GC-EID), high performance liquid chromatography (HPLC), ion chromatography (IC), and total organic carbon (TOC), the temporal sequence of phorate ozonation was proposed in this study. The oxidation of sulfur atoms on the phosphorus-sulfur double bond or carbon-sulfur-carbon bond by ozonation was found to occur at first to form sulfate and various intermediates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Phorat, PESTANAL®, analytical standard