Merck
  • Copper and zinc promote interactions between membrane-anchored peptides of the metal binding domain of the prion protein.
Copper and zinc promote interactions between membrane-anchored peptides of the metal binding domain of the prion protein.
Biochemistry (2007-03-21)
Angela G Kenward, Libero J Bartolotti, Colin S Burns
ZUSAMMENFASSUNG
The prion protein (PrP) has been identified as a metalloprotein capable of binding multiple copper ions and possibly zinc. Recent studies now indicate that prion self-recognition may be an important factor in both the normal function and misfunction of this protein. We have developed fluorescently labeled models of the prion protein that allow prion-prion interactions and metal binding to be investigated on the molecular level. Peptides encompassing the full metal binding region were anchored to the surface of small unilamellar vesicles, and PrP-PrP interactions were monitored by fluorescence spectroscopy as a function of added metal. Both Cu2+ and Zn2+ were found to cause an increase in the level of PrP-PrP interactions, by 117 and 300%, respectively, whereas other metals such as Ni2+, Co2+, and Ca2+ had no effect. The binding of either of these cofactors appears to act as a switch that induces PrP-PrP interactions in a reversible manner. Both glutamine and tryptophan residues, which occur frequently in the metal binding region of PrP, were found to be important in mediating PrP-PrP interactions. Experiments demonstrate that tryptophan residues are also responsible for the low level of PrP-PrP interactions observed in the absence of Cu2+ and Zn2+, and this is further supported by molecular modeling. Overall, our results indicate that PrP may be a bifunctional molecule capable of responding to fluctuations in both neuronal Cu2+ and Zn2+ levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Pyrenbuttersäure, 97%