Merck
  • Room temperature phosphorescence of alpha-bromonaphthalene induced by cyclodextrin in the presence of hexahydropyridine or 1-ethylpiperidine and its application.
Room temperature phosphorescence of alpha-bromonaphthalene induced by cyclodextrin in the presence of hexahydropyridine or 1-ethylpiperidine and its application.
Analytica chimica acta (2007-03-28)
Ya-Xian Zhu, Jing-He Peng, Yong Zhang
ZUSAMMENFASSUNG
Two novel heterocyclic third components, hexahydropyridine (HHP) and 1-ethylpiperidine (EP) were firstly found to enhance room temperature phosphorescence (RTP) of alpha-bromonaphthalene (alpha-BrN) induced by cyclodextrin. The effects of equilibrium time for formation of inclusion complex, temperature, pH values and the variation of concentrations of each component on RTP of alpha-BrN and the RTP lifetime of each ternary complex had been investigated and compared to discuss inclusion mechanism of ternary complexes. The RTP lifetimes of alpha-BrN/beta-CD/HHP, alpha-BrN/beta-CD/cyclohexane (CH) and alpha-BrN/beta-CD/EP were 6.18, 7.71 and 9.36 ms, respectively. Based on the strongest RTP of alpha-BrN induced by CD in the presence of EP, a method for determination of EP was established. Under the optimal conditions, the analytical curve of EP gave a liner dynamic range of 1.50x10(-4) to 1.50x10(-3) mol L(-1) with a detection limit of 4.8x10(-5) mol L(-1). When the established CD-RTP method was applied to determine the concentration of EP synthetic samples in distilled water, the experimental results demonstrated that the recovery was 91.4% with a relative standard deviation less than 2.85% (n=7).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Brom-naphthalin, 97%
Sigma-Aldrich
1-Ethylpiperidin, 99%
Sigma-Aldrich
1-Brom-naphthalin, ≥95%