Merck
Lithium diisopropylamide: solution kinetics and implications for organic synthesis.
Angewandte Chemie (International ed. in English) (2007-03-28)
David B Collum, Anne J McNeil, Antonio Ramirez
ZUSAMMENFASSUNG
Lithium diisopropylamide (LDA) is a prominent reagent used in organic synthesis. In this Review, rate studies of LDA-mediated reactions are placed in the broader context of organic synthesis in three distinct segments. The first section provides a tutorial on solution kinetics, emphasizing the characteristic rate behavior caused by dominant solvation and aggregation effects. The second section summarizes substrate- and solvent-dependent mechanisms that reveal basic principles of solvation and aggregation. The final section suggests how an understanding of mechanism might be combined with empirical methods to optimize yields, rates, and selectivities of organolithium reactions and applied to organic synthesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumdiisopropylamid -Lösung, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Lithiumdiisopropylamid -Lösung, 1.0 M in THF/hexanes
Sigma-Aldrich
Diisopropylamin, ≥99.5%
Sigma-Aldrich
Lithiumdiisopropylamid, 97%
Sigma-Aldrich
Diisopropylamin, purified by redistillation, 99.95%
Sigma-Aldrich
Diisopropylamin, puriss. p.a., ≥99.0% (GC)
Supelco
Diisopropylamin, analytical standard