Merck
  • A comparative study of micellar and microemulsion EKC for the analysis of benzoylurea insecticides and their analogs.
A comparative study of micellar and microemulsion EKC for the analysis of benzoylurea insecticides and their analogs.
Electrophoresis (2007-04-24)
Xia Yang, Yan Xia, Chuanjiang Tao, Yiping Liao, Yumin Zuo, Huwei Liu
ZUSAMMENFASSUNG
An investigation of the basic factors which govern the microemulsion EKC (MEEKC) and MEKC for the separation of four benzoylurea (BU) insecticides and their four analogs was carried out. In MEEKC, the separation of eight BU compounds was optimized by changing the microemulsion composition, such as concentration of SDS, octane, n-butanol, and isopropanol percentages, as well as capillary temperature. Separation optimization was also carried out for MEKC, showing that ACN and a high level of another additive gamma-CD were needed to achieve effective separation of these analytes. Although separation with baseline resolution was achieved by either MEEKC or MEKC methods, the separation selectivity resulting from the proposed MEEKC method was completely different from that of MEKC. In addition, analytical time in MEEKC was longer than that in MEKC, but in view of theoretical plate numbers, detection limits, and reproducibility, both methods were effective for the analysis of BU insecticides and their analogs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Flufenoxuron, PESTANAL®, analytical standard
Supelco
Hexaflumuron, PESTANAL®, analytical standard