Merck
Suppression of ochratoxin biosynthesis by naturally occurring alkaloids.
Food additives and contaminants (2007-04-25)
S E Lee, B S Park, P Bayman, J L Baker, W S Choi, B C Campbell
ZUSAMMENFASSUNG
The effects of four alkaloids on the biosynthesis of ochratoxin A (OTA), ochratoxin B (OTB) and citrinin were examined on four OTA-producing aspergilli: Aspergillus auricomus, A. sclerotiorum and two isolates of A. alliaceus. Piperine and piperlongumine, natural alkaloids of Piper longum, significantly inhibited OTA production at 0.001% (w/v) for all aspergilli examined. Piperine and piperlongumine affected the polyketide synthesis step of OTA production and inhibited production of citrinin. Curcumin, a constituent of tumeric, completely inhibited mycelial growth of A. alliaceus isolate 791 at 0.1% (w/v) and decreased OTA production by approximately 70% at 0.01% (w/v). Sesamin, a constituent of sesame oil, inhibited OTA and OTB production by 60 and 45%, respectively, at 0.1% (w/v), showing its effect was on chloroperoxidase and polyketide synthase activity. The potential advantage of these natural products to reduce ochratoxin contamination of agricultural commodities is discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ochratoxin B -Lösung, ~10 μg/mL in acetonitrile, analytical standard