Merck
The responses of rainbow trout gills to high lithium and potassium concentrations in water.
Ecotoxicology and environmental safety (2007-05-19)
Victoria Tkatcheva, Ismo J Holopainen, Heikki Hyvärinen, Jussi V K Kukkonen
ZUSAMMENFASSUNG
The objective of this study is to discern the effects of increased waterborne lithium and potassium on rainbow trout gill histology, lipid composition, and enzyme activity. The study aims to elucidate the effects of these ions in the laboratory in concentrations similar to those prevailing in a forest lake Poppalijärvi in a contaminated mining area in NW Russia. Under the lithium and potassium exposure, the fish were further stressed by high pH (8.2 as in Lake Poppalijärvi) and lack of food. These multiple stress conditions altered the gill membrane fluidity by increasing sphingomyelin (5.5+/-0.6 compared to 2.9+/-0.3% in the control) and reducing cholesterol (4+/-1 compared to 17+/-3 mg g(-1) in the control). The total ATPase activity tended to be higher in the lithium-potassium-exposed group (46+/-6 compared to 34+/-2 micromol Pi h(-1) mg(-1)protein in the control). Lithium toxicity was lowered here by the protective role of higher potassium contents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumsulfat, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Kaliumsulfat, ACS reagent, ≥99.0%, powder or crystals
Sigma-Aldrich
Kaliumbisulfat, reagent grade
Sigma-Aldrich
Kaliumbisulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Kaliumsulfat, 99.99% trace metals basis
Sigma-Aldrich
Kaliumsulfat, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Kaliumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumsulfat, suitable for plant cell culture