Merck
  • Analysis of alternariol and alternariol monomethyl ether on flavedo and albedo tissues of tangerines (Citrus reticulata) with symptoms of alternaria brown spot.
Analysis of alternariol and alternariol monomethyl ether on flavedo and albedo tissues of tangerines (Citrus reticulata) with symptoms of alternaria brown spot.
Journal of agricultural and food chemistry (2007-06-01)
Rodrigo F Magnani, Gezimar D De Souza, Edson Rodrigues-Filho
ZUSAMMENFASSUNG
A method was developed for the quantification of alternariol and alternariol monomethyl ether on tangerines with and without symptoms of Alternaria brown spot disease. The method employs solid-phase extraction for cleanup, followed by high-performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS/MS) for detection. This method was validated on flavedo (exocarp or epicarp, exterior yellow peel) and on albedo tissue (mesocarp, interior white peel). An excellent linearity over a range of 0.50-20.0 mg/kg was achieved, with r2 >or= 0.997. The limits of detection (LOD) and quantification (LOQ) were fewer than 0.13 and 0.50 microg/kg, respectively. The relative standard deviations (RSDs) were <or=14.4% during the validation. The levels of these mycotoxins on flavedo of fruits with symptoms vary from 2.54 +/- 0.24 to 17.40 +/- 1.05 mug/kg. Surprisingly, neither alternariol nor altenariol monomethyl ether was detected on albedo tissues, suggesting that flavedo works as a barrier for such substances.