- Chromosome aberration test of Pigment Yellow 34 (lead chromate) in Chinese hamster ovary cells.
Chromosome aberration test of Pigment Yellow 34 (lead chromate) in Chinese hamster ovary cells.
Mutation research (2007-07-24)
Earle R Nestmann, Baowei Zhang
PMID17644466
ZUSAMMENFASSUNG
Lead chromate pigment in the form of the commercial pigment, Pigment Yellow 34, CAS No. 1344-37-2, used in the plastics and coatings industries, did not induce chromosome aberrations in Chinese hamster ovary (CHO) cell line WB(L). Lead chromate pigment is essentially insoluble in water, and in an effort to test the material under realistic conditions, no attempt to solubilize the pigment was made. These results are significant because others have reported lead chromate to cause genotoxicity in various assays, but only under conditions in which its aqueous solubility was artificially enhanced.