Merck
Cellular effects of some metabolic oxidation products pertinent to 4-ethoxyaniline.
Pharmacology & toxicology (1991-08-01)
T Lindqvist, P Moldéus, B Lindeke
ZUSAMMENFASSUNG
The toxicity of some metabolic products pertinent to 4-ethoxyaniline in isolated hepatocytes were investigated. The compounds investigated were 4-ethoxynitrosobenzene (1), 4-ethoxy-4'-nitrosodiphenylamine (2), 3,6-bis(4-ethoxy-phenylimino)-4-ethoxy-1,4-cyclohexadienylamine (3), 4-(4-ethoxyphenylimino)-2,3-dimethyl-2,5-cyclohexadiene-1-one (4) and 4-(4-ethoxyphenylimino)-2,6-dimethyl-2,5-cyclohexadiene-1-one (5). Of these, 1, 2 and 3 are oxidation products of 4-ethoxyaniline. Compounds 4 and 5 are dimethyl analogues of previously investigated oxidation product 4-(4-ethoxyphenylimino(-2,5-cyclohexadiene-1-one (NEPBQI). Among the investigated compounds, 1 and 2 were the most toxic towards isolated hepatocytes. In hepatocytes treated with compounds 1, 2 and 4, loss of cell viability was also accompanied by surface bleb formation. All compounds except 3 reacted with GSH resulting in depletion of cellular GSH. No formation of GSSG was observed, however. Thus, the GSH depletion was apparently due to conjugate formation rather than oxidation. No superoxide dismutase inhibitable reduction of acetylated cytochrome c was observed, thus none of the compounds undergoes measurable redox cycling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Ethoxyanilin, 98%