Merck
  • Cyanopyridyl containing 1,4-dihydroindeno[1,2-c]pyrazoles as potent checkpoint kinase 1 inhibitors: improving oral biovailability.
Cyanopyridyl containing 1,4-dihydroindeno[1,2-c]pyrazoles as potent checkpoint kinase 1 inhibitors: improving oral biovailability.
Bioorganic & medicinal chemistry letters (2007-09-05)
Yunsong Tong, Magdalena Przytulinska, Zhi-Fu Tao, Jennifer Bouska, Kent D Stewart, Chang Park, Gaoquan Li, Akiyo Claiborne, Peter Kovar, Zehan Chen, Philip J Merta, Mai-Ha Bui, Amanda Olson, Donald Osterling, Haiying Zhang, Hing L Sham, Saul H Rosenberg, Thomas J Sowin, Nan-Horng Lin
ZUSAMMENFASSUNG
A series of 1,4-dihydroindeno[1,2-c]pyrazole compounds with a cyanopyridine moiety at the 3-position of the tricyclic pyrazole core was explored as potent CHK-1 inhibitors. The impact of substitutions at the 6 and/or 7-position of the core on pharmacokinetic properties was studied in detail. Compounds carrying a side chain with an ether linker at the 7-position and a terminal morpholino group, such as 29 and 30, exhibited much-improved oral biovailability in mice as compared to earlier generation inhibitors. These compounds also possessed desirable cellular activity in potentiating doxorubicin and will serve as valuable tool compounds for in vivo evaluation of CHK-1 inhibitors to sensitize DNA-damaging agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Inden, ≥99%
Sigma-Aldrich
Inden, 98%
Sigma-Aldrich
Inden, contains 50-100 ppm tert-butylcatechol as stabilizer, technical grade, ≥90%