Merck
Salbutamol, terbutaline and pirbuterol allergy in an asthmatic patient.
Allergy (2007-09-12)
P Bonniaud, N Favrolt, E Collet, J-P Dumas, L Guilloux, G Pauli, P Camus
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Pirbuterol-acetat, VETRANAL®, analytical standard