Merck
  • A new strategy for the synthesis of benzylic sulfonamides: palladium-catalyzed arylation and sulfonamide metathesis.
A new strategy for the synthesis of benzylic sulfonamides: palladium-catalyzed arylation and sulfonamide metathesis.
The Journal of organic chemistry (2007-09-21)
Jonathan B Grimm, Matthew H Katcher, David J Witter, Alan B Northrup
ZUSAMMENFASSUNG
An efficient two-step strategy has been developed to access diversely functionalized benzylic sulfonamides. Execution of this strategy required the development of two reaction methods: the palladium-catalyzed cross-coupling of aryl halides with CH-acidic methanesulfonamides and a metathesis reaction between the resulting alpha-arylated sulfonamides and diverse amines. The broad scope of the cross-coupling process combined with a versatile sulfonamide metathesis constitutes an efficient strategy for the synthesis of various benzylic sulfonamides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methansulfonamid, ≥97.0% (CHN)