Merck
  • Estimating half-lives of pesticides in/on vegetation for use in multimedia fate and exposure models.
Estimating half-lives of pesticides in/on vegetation for use in multimedia fate and exposure models.
Chemosphere (2007-10-09)
Ronnie Juraske, Assumpció Antón, Francesc Castells
ZUSAMMENFASSUNG
Degradation half-lives in/on vegetation are needed in environmental risk assessment of pesticides, but these data are often not available for most active ingredients. To address this, we first correlated experimental soil degradation half-life data of 41 pesticides obtained from the reviewed literature with the corresponding experimental half-lives on plant surface. Degradation half-lives in soil were found to be four times slower compared with half-lives on plant surfaces. In a second step, we explored measured plant surface half-lives directly with those in vegetation. The results were validated by comparing computed values with results obtained from an experimental set-up. The uptake and dissipation of alpha-cypermethrin (insecticide) and bromopropylate (acaricide) was studied by detecting pesticide residues in whole and peeled tomato fruits using gas chromatography. Half-lives within vegetation were found to be four times faster compared with plant surface half-lives. Using this experimental based approach, it is concluded that the estimation of degradation half-lives of pesticides in/on vegetation to be used as input data in environmental mass balance models can be directly correlated from the more abundant ready experimental degradation half-life data for soil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Bromopropylat, PESTANAL®, analytical standard