Merck
Catalytic oxidation of hydrazo derivatives promoted by a TiCl3/HBr system.
Journal of the American Chemical Society (2007-10-26)
Eyal Drug, Michael Gozin
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Titan(III)-chlorid -Lösung, 10-15% TiCl3 basis
Sigma-Aldrich
Titan(III)-chlorid, ≥99.995% trace metals basis