Merck
  • 5-Furan-2yl[1,3,4]oxadiazole-2-thiol, 5-furan-2yl-4H [1,2,4] triazole-3-thiol and their thiol-thione tautomerism.
5-Furan-2yl[1,3,4]oxadiazole-2-thiol, 5-furan-2yl-4H [1,2,4] triazole-3-thiol and their thiol-thione tautomerism.
Molecules (Basel, Switzerland) (2007-11-17)
M Koparir, A Cetin, A Cansiz
ZUSAMMENFASSUNG
5-Furan-2-yl[1,3,4]oxadiazole-2-thiol (Ia) and 5-furan-2-yl-4H-[1,2,4]-triazole-3-thiol (Ib) were synthesized from furan-2-carboxylic acid hydrazide. Mannich bases and methyl derivatives were then prepared. The structures of the synthesized compounds were confirmed by elemental analyses, IR and (1)H-NMR spectra. Their thiol-thione tautomeric equilibrium is described.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Furosäure, 98%