- Polymer-supported Cinchona alkaloid-derived ammonium salts as recoverable phase-transfer catalysts for the asymmetric synthesis of alpha-amino acids.
Polymer-supported Cinchona alkaloid-derived ammonium salts as recoverable phase-transfer catalysts for the asymmetric synthesis of alpha-amino acids.
Molecules (Basel, Switzerland) (2007-11-17)
Rafael Chinchilla, Patricia Mazón, Carmen Nájera
PMID18007436
ZUSAMMENFASSUNG
Alkaloids such as cinchonidine, quinine and N-methylephedrine have been N-alkylated using polymeric benzyl halides or co-polymerized and then N-alkylated, thus affording a series of polymer-supported chiral ammonium salts which have been employed as phase-transfer catalysts in the asymmetric benzylation of an N-(diphenylmethylene)glycine ester. These new polymeric catalysts can be easily recovered by simple filtration after the reaction and reused. The best ee's were achieved when Merrifield resin-anchored cinchonidinium ammonium salts were employed.
MATERIALIEN