Merck
  • The analysis and application of a recombinant monooxygenase library as a biocatalyst for the Baeyer-Villiger reaction.
The analysis and application of a recombinant monooxygenase library as a biocatalyst for the Baeyer-Villiger reaction.
Journal of microbiology and biotechnology (2007-12-07)
Jiyeoun Park, Donghyun Kim, Sujin Kim, Jinhee Kim, Kihwan Bae, Choonghwan Lee
ZUSAMMENFASSUNG
Because of their selectivity and catalytic efficiency, BVMOs are highly valuable biocatalysts for the chemoenzymatic synthesis of a broad range of useful compounds. In this study, we investigated the microbial Baeyer-Villiger oxidation and sulfoxidation of thioanisole and bicyclo[3.2.0]hept-2-en-6-one using whole Escherichia coli cells that recombined with each of the Baeyer-Villiger monooxygenases originated from Pseudomonas aeruginosa PAO1 and two from Streptomyces coelicolor A3(2). The three BVMOs were identified in the microbial genome database by a recently described protein sequence motif; e.g., BVMO motif (FXGXXXHXXXW). The reaction products were identified as (R)-/(S)sulfoxide and 2-oxabicyclo/3-oxabicyclo[3.3.0]oct-6-en-2-one by GC-MS analysis. Consequently, this study demonstrated that the three enzymes can indeed catalyze the Baeyer-Villiger reaction as a biocatalyst, and effective annotation tools can be efficiently exploited as a source of novel BVMOs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thioanisol, ReagentPlus®, ≥99%