- Purification and characterization of 2-aminoacetophenone reductase of newly isolated Burkholderia sp. YT.
Purification and characterization of 2-aminoacetophenone reductase of newly isolated Burkholderia sp. YT.
Journal of bioscience and bioengineering (2007-12-19)
Keiko Yamada-Onodera, Yuhki Takase, Yoshiki Tani
PMID18086443
ZUSAMMENFASSUNG
We found that a newly isolated Burkholderia sp. produced (R)-2-amino-1-phenylethanol from 2-aminoacetophenone, showing the high stereospecificity. NADPH-dependent 2-aminoacetophenone reductase purified to homogeneity was a dimer with a molecular mass of 65,000. The purified enzyme did not reduce acetophenone and 1-phenyl-1-propanone. The purified enzyme converted 2-aminoacetophenone to only (R)-2-amino-1-phenylethanol.