Merck
  • Effects of CYP inducers and inhibitors on the pharmacokinetics of intravenous theophylline in rats: involvement of CYP1A1/2 in the formation of 1,3-DMU.
Effects of CYP inducers and inhibitors on the pharmacokinetics of intravenous theophylline in rats: involvement of CYP1A1/2 in the formation of 1,3-DMU.
The Journal of pharmacy and pharmacology (2007-12-20)
Kyung H Yang, Joo H Lee, Myung G Lee
ZUSAMMENFASSUNG
The types of hepatic cytochrome P450 (CYP) isozymes responsible for the metabolism of theophylline and for the formation of 1,3-dimethyluric acid (1,3-DMU) in rats in-vivo does not seem to have been studied at the dose ranges of dose-independent metabolic disposition of theophylline in rats (up to 10 mg kg(-1)). Therefore, theophylline (5 mg kg(-1)) was administered i.v. to male Sprague-Dawley rats pretreated with various inducers and inhibitors of CYP isozymes. In rats pretreated with 3-methylcholanthrene (3-MC), orphenadrine or dexamethasone (main inducers of CYP1A1/2, CYP2B1/2 and CYP3A1/2, respectively, in rats), the time-averaged non-renal clearance (CLNR) of theophylline was significantly faster than in their respective controls (1260, 42.7 and 69.0% increases, respectively). However, in rats pretreated with troleandomycin (a major inhibitor of CYP3A1/2 in rats), CLNR was significantly slower than in the controls (50.7% decrease). The 24 h urinary excretion of 1,3-DMU was increased significantly only in rats pretreated with 3-MC. The ratio of area under the curve for 1,3-DMU and theophylline (AUC1,3-DMU/AUCtheophylline) was increased significantly in rats pretreated with 3-MC (160% increase) and decreased significantly in rats pretreated with troleandomycin (50.1% decrease); however, the ratio was not increased in rats pretreated with dexamethasone. These data suggest that theophylline is primarily metabolized via CYP1A1/2, CYP2B1/2, and CYP3A1/2, and that 1,3-DMU is primarily formed via CYP1A1/2, and possibly CYP3A1/2, in rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Orphenadrin -hydrochlorid, ≥98.0% (AT)
Supelco
Orphenadrin -citrat (Salz), analytical standard
Orphenadrin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard