Merck
  • Synthesis and quantitative evaluation of Glycero-D-manno-heptose binding to concanavalin A by fluorous-tag assistance.
Synthesis and quantitative evaluation of Glycero-D-manno-heptose binding to concanavalin A by fluorous-tag assistance.
Angewandte Chemie (International ed. in English) (2008-01-19)
Firoz A Jaipuri, Beatrice Y M Collet, Nicola L Pohl
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimethylsilyltrifluormethansulfonat, 99%
Sigma-Aldrich
Trimethylsilyltrifluormethansulfonat, purum, ≥98.0% (T)