Merck
  • High levels of saturated very long-chain fatty acid (hexacosanoic acid; C26:0) in whole blood are associated with metabolic syndrome in Japanese men.
High levels of saturated very long-chain fatty acid (hexacosanoic acid; C26:0) in whole blood are associated with metabolic syndrome in Japanese men.
Diabetes research and clinical practice (2008-02-05)
Atsumi Kume, Tetsuro Miyazaki, Yohei Kitamura, Kyoichi Oshida, Naotake Yanagisawab, Hirotaka Takizawa, Kiyotaka Fujii, Takashi Kiyanagi, Katsuhiko Sumiyoshi, Hirotoshi Ohmura, Hiroshi Mokuno, Kazunori Shimada, Hiroyuki Daida
ZUSAMMENFASSUNG
Hexacosanoic acid (C26:0) is a saturated very long-chain fatty acid and high levels of C26:0 in red blood cells are reported to be closely related with risk factors of atherosclerosis. However, the relationship between absolute levels of C26:0 in whole blood and metabolic syndrome (MS) has not been determined. We divided 218 consecutive apparently healthy male subjects into an MS group (n=78) and a non-MS group (n=140) according to the definition of the International Diabetes Federation. The levels of C26:0 in whole blood were measured by gas liquid chromatography-mass spectrometry. The MS group had significantly higher levels of C26:0 than the non-MS group (2.42+/-0.31mug/ml vs. 2.25+/-0.29mug/ml, P=0001). There was a significant association between the levels of C26:0 and the number of factors of MS. The levels of C26:0 positively correlated with age, blood pressure, triglyceride and fasting plasma glucose. Multivariate analysis revealed that the level of C26:0 is still an independent variable for the presence of MS after adjustment for age and each criterion of MS. The absolute levels of C26:0 in whole blood appear to be associated with MS independent of its component parts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexacosansäure, ≥95% (capillary GC)
Sigma-Aldrich
Hexacosansäure, technical, ≥90% (GC)