Merck
  • Synthesis and structure-activity relationships of the first ferrocenyl-aryl-hydantoin derivatives of the nonsteroidal antiandrogen nilutamide.
Synthesis and structure-activity relationships of the first ferrocenyl-aryl-hydantoin derivatives of the nonsteroidal antiandrogen nilutamide.
Journal of medicinal chemistry (2008-02-29)
Olivier Payen, Siden Top, Anne Vessières, Emilie Brulé, Marie-Aude Plamont, Michael J McGlinchey, Helge Müller-Bunz, Gérard Jaouen
ZUSAMMENFASSUNG
We present here the first synthesis of organometallic complexes derived from the nonsteroidal antiandrogen nilutamide, bearing a ferrocenyl substituent at position N(1) or at C(5) of the hydantoin ring; for comparison, we also describe the C(5) p-anisyl organic analogue. All of these complexes retain a modest affinity for the androgen receptor. The N-substituted complexes show a weak or moderate antiproliferative effect (IC 50 around 68 microM) on hormone-dependent and -independent prostate cancer cells, while the C(5)-substituted compounds exhibit toxicity levels 10 times higher (IC 50 around 5.4 microM). This strong antiproliferative effect is probably due to a structural effect linked to the aromatic character of the ferrocene rather than to its organometallic feature. In addition, it seems connected to a cytotoxic effect rather than an antihormonal one. These results open the way toward a new family of molecules that are active against both hormone-dependent and hormone-independent prostate cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nilutamid, solid
Nilutamid, European Pharmacopoeia (EP) Reference Standard