Merck
Betablockers at work: the crystal structure of the beta2-adrenergic receptor.
Angewandte Chemie (International ed. in English) (2008-03-19)
Felix Hausch
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Carazolol, analytical standard