Merck
  • Enhanced depigmenting effects of N-glycosylation inhibitors delivered by pH-sensitive liposomes into HM3KO melanoma cells.
Enhanced depigmenting effects of N-glycosylation inhibitors delivered by pH-sensitive liposomes into HM3KO melanoma cells.
Journal of cosmetic science (2008-04-15)
Ju Young Park, Hyunjung Choi, Jae Sung Hwang, Junoh Kim, Ih-Seop Chang
ZUSAMMENFASSUNG
Delivery activity of pH-sensitive 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE):cholesteryl hemisuccinate (CHEMS) liposomes was assessed as an in vitro intracellular carrier system to increase the bioavailability of depigmentation actives. N-glycosylation inhibitors have a glycosylation-inhibiting effect, which is useful for the skin depigmentation that operates by interfering with the maturation of tyrosinase. However, an N-glycosylation inhibitor does not easily pass through skin or even cellular membranes due to its water-soluble property. Therefore, it should be transported to target cells by an efficient delivery carrier to reduce the glycosylated tyrosinase. Glycosylation-inhibiting and depigmentation effects of N-butyldeoxynojirimycine (NB-DNJ) and 1-deoxynojirimycine (DNJ)-loaded liposomes were evaluated using Western blotting and measurement of synthesized melanin. Interestingly, it was found that the pH-sensitive liposomes increased the glycosylation-inhibiting and thus, pigment-lightening effects of N-glycosylation inhibitors in vitro. In addition, cargo materials loaded in pH-sensitive liposomes were found to be much more efficiently delivered into the cytoplasm, as observed in fluorescent-activated cell sorting (FACS) and confocal laser-scanning microscopic (CLSM) analysis. These results indicate that pH-sensitive DOPE:CHEMS liposomes have a strong potential as a carrier system to promote delivery efficiency and to enhance the biological effects of water-soluble actives for applications in cosmetics, personal care products, and pharmaceutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterylhemisuccinat