Merck
  • Chlorination and thermal degradation of 2-chlorodibenzodioxin and dibenzofuran by CuCl2 or CuCl at 350 degrees C.
Chlorination and thermal degradation of 2-chlorodibenzodioxin and dibenzofuran by CuCl2 or CuCl at 350 degrees C.
Chemosphere (2008-04-29)
Nicolas Visez, Jean-Pierre Sawerysyn
ZUSAMMENFASSUNG
Chlorination and thermal degradation of 2-chlorodibenzodioxin and dibenzofuran were investigated using sealed tubes at 350 degrees C in the presence of CuCl(2) or CuCl. Twelve organic species, including polychlorobenzenes, were identified and quantified as a function of residence time. Time behaviours of PCDDs and PCDFs regarding destruction processes were compared. The formation of polychlorobenzenes from the thermal heterogeneous degradation of PCDDs was only a minor pathway but it could contribute to the good linear correlation observed between the concentration of polychlorobenzenes and those of PCDD/Fs in the emissions of municipal solid waste incinerators.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(I)-chlorid, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Kupfer(I)-chlorid, ≥99.995% trace metals basis
Sigma-Aldrich
Kupfer(I)-chlorid, AnhydroBeads, ≥99.99% trace metals basis