Merck
  • Mequinol 2%/tretinoin 0.01% topical solution for the treatment of melasma in men: a case series and review of the literature.
Mequinol 2%/tretinoin 0.01% topical solution for the treatment of melasma in men: a case series and review of the literature.
Cutis (2008-04-30)
Jon Keeling, Lina Cardona, Adolpho Benitez, Rachel Epstein, Marta Rendon
ZUSAMMENFASSUNG
Melasma is a common hyperpigmentation disorder that typically affects women, though up to 10% of white individuals seeking treatment for melasma are men. Melasma can be a source of embarrassment for men because of its association with women and pregnancy. We performed a case series assessing the use of mequinol 2%/ tretinoin 0.01% topical solution in 5 men with melasma. Four of 5 patients achieved complete clearance of melasma at 12 weeks, and 1 patient showed moderate improvement. Side effects were minimal and consisted of stinging in one patient. All patients maintained results at the 16-week follow-up visit. Mequinol 2%/tretinoin 0.01% topical solution was an effective and well-tolerated treatment of melasma in men. The vehicle resulted in good compliance and minimal adverse effects in patients. This is the first report describing the use of mequinol 2%/tretinoin 0.01% topical solution for the treatment of melasma in men; there are no reports in women.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
4-Methoxyphenol, ReagentPlus®, 99%
Supelco
Retinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Methoxyphenol, purum, ≥98.0% (HPLC)
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard