Merck
Liquid phase separation of 1-butene from 2-butenes on all-silica zeolite RUB-41.
Chemical communications (Cambridge, England) (2008-05-21)
Bart Tijsebaert, Csaba Varszegi, Hermann Gies, Feng-Shou Xiao, Xinhe Bao, Takashi Tatsumi, Ulrich Müller, Dirk De Vos
ZUSAMMENFASSUNG
The all-silica zeolite RUB-41, containing 8- and 10-membered rings, is able to separate trans-2-butene and cis-2-butene from 1-butene and represents a possible improvement in isolating pure 1-butene from a butene mixture.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Buten, ≥99%