Merck
  • Synthesis and identification of hydroxylated metabolites of the anti-estrogenic agent cyclofenil.
Synthesis and identification of hydroxylated metabolites of the anti-estrogenic agent cyclofenil.
Journal of mass spectrometry : JMS (2008-06-26)
Peter Gärtner, Karin Hofbauer, Christian Reichel, Thomas Geisendorfer, Günter Gmeiner
ZUSAMMENFASSUNG
The detection of metabolites of the anti-estrogenic substance cyclofenil, listed on the World Anti-Doping Agency (WADA) Prohibited List since 2004 is described. Target substances are hydroxylated metabolites, bearing an aliphatic hydroxyl group either in the 2-, 3- or 4-position of the aliphatic ring, in addition to the phenolic functions on the aromatic rings. Structural identification used NMR as well as high-resolution mass spectrometry after nano-electrospray ionisation (ESI). Unambiguous detection of all three synthesised cyclofenil metabolites M1-M3 was done using gas chromatography for separation and electron ionisation mass spectrometry for detection of the per-silylated compounds in comparison with a reference urine deriving from an excretion study within the WADA 2007 Educational Programme.