Merck
  • Determination of tetracyclines residues in honey by on-line solid-phase extraction high-performance liquid chromatography.
Determination of tetracyclines residues in honey by on-line solid-phase extraction high-performance liquid chromatography.
Talanta (2008-07-01)
Jiantao Li, Ligang Chen, Xiao Wang, Haiyan Jin, Lan Ding, Kun Zhang, Hanqi Zhang
ZUSAMMENFASSUNG
An automated system using on-line solid-phase extraction (SPE) high-performance liquid chromatography (HPLC) with ultraviolet (UV) detection was developed for the determination of tetracyclines (TCs), such as tetracycline (TC), oxytetracycline (OTC), chlortetracycline (CTC), metacycline (MC), and doxycycline (DC) in honey. One milliliter diluted honey sample was injected into a conditioned C18 SPE column and the matrix was washed out with water for 3 min. By rotation of the switching valve, TCs were eluted and transferred to the analytical column by the chromatographic mobile phase. Chromatographic conditions were optimized. TCs were separated in less than 8 min with a gradient elution using a mixture of 0.8% formic acid and acetonitrile. The UV detection was performed at 365 nm. The conditions for on-line SPE, including solvent and total time for loading sample and washing matrix were also optimized. Time for extraction and separation decreased greatly. For the five kinds of TCs, the limits of detection (LODs) at a signal-to-noise of 3 ranged from 5 to 12 ng g(-1). The relative standard deviations (R.S.D.) for the determination of TCs ranged from 3.4 to 7.1% within a day and ranged from 3.2 to 8.9% in 3 days, respectively.