Merck
Stereochemistry and rearrangement reactions of hydroxylignanolactones.
Organic & biomolecular chemistry (2008-07-05)
Barbara Raffaelli, Monika Pohjoispää, Tapio Hase, Christine J Cardin, Yu Gan, Kristiina Wähälä
ZUSAMMENFASSUNG
Various conflicting data on the rearrangement and absolute stereochemistry of hydroxylignano-9,7'-lactones are resolved using 18O labeled compounds, also confirmed by an X-ray analysis of a pure lignano-9,7'-lactone enantiomer, obtained for the first time. Under NaH/DMF rearrangement conditions a silyl protected hydroxylignano-9,9'-lactone underwent an unexpected silyl migration.