Merck
  • [Study on chemical constituents and anti-bacterial activity of volatile oil from seed of Aquilaria sinensis].
[Study on chemical constituents and anti-bacterial activity of volatile oil from seed of Aquilaria sinensis].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2008-07-16)
Jun Liu, Wen-Li Mei, Hai-Bin Cui, Jiao Wu, Hao-Fu Dai
ZUSAMMENFASSUNG
Volatile oil from the seed of Aquilaria sinensis was extracted with Petroleum ether. Anti-bacterial bioassay of the oil against methicillin-resistant Staphylococcus aureus (MRSA) was carried out by paper disco diffusion method. The result showed that the oil possessed inhibitory activity towards MRSA. 19 compounds of the volatile oil were identified by GC-MS analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyllinoleat, ≥99%
Sigma-Aldrich
Ethyllinoleat, technical, ≥65% (GC)