- Sterically biased 3,3-sigmatropic rearrangement of chiral allylic azides: application to the total syntheses of alkaloids.
Sterically biased 3,3-sigmatropic rearrangement of chiral allylic azides: application to the total syntheses of alkaloids.
The Journal of organic chemistry (2008-07-23)
Sophie Lauzon, François Tremblay, David Gagnon, Cédrickx Godbout, Christine Chabot, Catherine Mercier-Shanks, Stéphane Perreault, Hélène DeSève, Claude Spino
PMID18642871
ZUSAMMENFASSUNG
We describe a tandem Mitsunobu/3,3-sigmatropic rearrangement of allylic azides on a chiral auxiliary system that favors one regioisomer thanks to its exceptional steric bias. The sequence may be completed by the oxidative cleavage of the auxiliary or by a ring-closing metathesis reaction that produces a carbo- or heterocycle directly and a recyclable form of the chiral auxiliary. Applications of the methodology to the total synthesis of (+)-coniine, (+)-lentiginosin, and (+)-pumiliotoxin C are reported.