- Analysis of urinary aromatic acids by liquid chromatography tandem mass spectrometry.
Analysis of urinary aromatic acids by liquid chromatography tandem mass spectrometry.
Biomedical chromatography : BMC (2008-07-26)
Brian Crow, Michael Bishop, Ekaterina Paliakov, Dean Norton, Joe George, J Alexander Bralley
PMID18655225
ZUSAMMENFASSUNG
The separation and detection of 11 urinary aromatic acids was developed using HPLC-MS/MS. The method features a simple sample preparation involving a single-step dilution with internal standard and a rapid 8 min chromatographic separation. The accuracy was evaluated by the recovery of known spikes between 87 and 110%. Inter- and intra-assay precision (CV) was below 11% in all cases and the analytes were observed to be stable for up to 8 weeks when stored at -20 degrees C. The method was validated based upon linearity, accuracy, precision and stability and was used to establish reference intervals for children and adults.
MATERIALIEN