Merck
  • Absolute rate constants for some intermolecular reactions of alpha-aminoalkylperoxyl radicals. Comparison with alkylperoxyls.
Absolute rate constants for some intermolecular reactions of alpha-aminoalkylperoxyl radicals. Comparison with alkylperoxyls.
The Journal of organic chemistry (2008-07-31)
Jacques Lalevée, Xavier Allonas, Jean-Pierre Fouassier, K U Ingold
ZUSAMMENFASSUNG
Seven alpha-aminoalkylperoxyl radicals have been generated by 355 nm laser flash photolysis (LFP) of oxygen-saturated di-tert-butyl peroxide containing mono-, di-, and trialkylamines and a dialkylarylamine. All these peroxyls possess absorptions in the near-UV (strongest for the trialkylamine-derived peroxyls) which permits direct monitoring of the kinetics of their reactions with many substrates. The measured rate constants for hydrogen atom abstraction from some phenols and oxygen atom transfer to triphenylphosphine demonstrated that all seven alpha-aminoalkylperoxyls have similar reactivities toward each specific substrate. More importantly, a comparison with literature data for alkylperoxyls shows that alpha-aminoalkylperoxyls and these alkylperoxyls have essentially the same reactivities. The combination of LFP and alkylamines provides a quick, reliable method for determining absolute rate constants for alkylperoxyl radical reactions, an otherwise laborious task.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Luperox® DI, tert-Butylperoxid, 98%