- Mucosal tear in collagenous colitis.
Mucosal tear in collagenous colitis.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association (2008-08-12)
Thomas Dunzendorfer, Sarah Wilkins, Rebecca Johnson
PMID18691941