Merck
  • Apatite formation abilities and mechanical properties of hydroxyethylmethacrylate-based organic-inorganic hybrids incorporated with sulfonic groups and calcium ions.
Apatite formation abilities and mechanical properties of hydroxyethylmethacrylate-based organic-inorganic hybrids incorporated with sulfonic groups and calcium ions.
Journal of materials science. Materials in medicine (2008-08-16)
Toshiki Miyazaki, Moriyoshi Imamura, Eiichi Ishida, Masahiro Ashizuka, Chikara Ohtsuki
ZUSAMMENFASSUNG
Apatite formation in the living body is an essential requirement for artificial materials to exhibit bone-bonding bioactivity. It has been recently revealed that sulfonic groups trigger apatite nucleation in a body environment. Organic-inorganic hybrids consisting of organic polymers and the sulfonic groups are therefore expected to be useful for preparation of novel bone-repairing materials exhibiting flexibility as well as bioactivity. In the present study, organic-inorganic hybrids were prepared from hydroxyethylmethacrylate (HEMA) in the presence of vinylsulfonic acid sodium salt (VSAS) and calcium chloride (CaCl(2)). The bioactivities of the hybrids were assessed in vitro by examining the apatite formation in simulated body fluid (SBF, Kokubo solution). The hybrids deposited on the apatite after soaking in SBF within 7 days. Tensile strength measurements showed a tendency to increase with increases in VSAS and CaCl(2) content. It was assumed that this phenomenon was attributed to the formation of cross-linking in the hybrids.