Merck
  • Yeast ribosomal proteins. VIII. Isolation of two proteins and sequence characterization of twenty-four proteins from cytoplasmic ribosomes.
Yeast ribosomal proteins. VIII. Isolation of two proteins and sequence characterization of twenty-four proteins from cytoplasmic ribosomes.
Molecular & general genetics : MGG (1984-01-01)
E Otaka, K Higo, T Itoh
ZUSAMMENFASSUNG
Two proteins, YL41 and YL43, were isolated from 80S ribosomes of Saccharomyces cerevisiae by filtration through a Sephacryl S-200 column and by chromatography on a column of carboxymethylcellulose. Their amino acid compositions are presented. Twenty-four proteins including these two proteins were subjected to sequence analyses by automated Edman degradation. Amino-terminal amino acid sequences were determined for 17 proteins,YS3, YS9, YS23, YS24, YS29, YL6, YL8, YLll, YLI5,YL17, YL23, YL28, YL33, YL37, YL39, YL41, and YL43.YL41, which has a 72.7% lysine and arginine content, was found to be particular to eukaryotic ribosomes. The amino-termini of another seven proteins, YS2, YS5, YS8, YS12,YS13, YS20, and YS27, were suggested to be blocked. Comparison of the amino-terminal sequences with all other ribosomal protein sequences so far available indicates that YS9 shows sequence homology to rat liver ribosomal protein S8 (Wittmann-Liebold et al. 1979).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sephacryl®, 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 400-HR, MW range 20-8000 kDa (globular proteins)
Sigma-Aldrich
Sephacryl®, 100-HR, MW range 1000-100,000 Da (globular proteins)
Sigma-Aldrich
Sephacryl®, 500-HR, MW range 40-20,000 kDa (dextrans)