Merck
  • Ring-enlargement of dimethylaminopropenoyl cyclopropanes: an efficient route to substituted 2,3-dihydrofurans.
Ring-enlargement of dimethylaminopropenoyl cyclopropanes: an efficient route to substituted 2,3-dihydrofurans.
The Journal of organic chemistry (2008-09-19)
Rui Zhang, Yongjiu Liang, Guangyuan Zhou, Kewei Wang, Dewen Dong
ZUSAMMENFASSUNG
A convenient and efficient synthesis of substituted dihydrofurans is developed via ring-enlargement of 1-dimethylaminopropenoyl-1-carbamoyl/benzoyl cyclopropanes catalyzed by ammonium acetate in acetic acid with high regio- and stereoselectivity. Some of the newly synthesized substituted dihydrofurans are subjected to further synthetic transformation in the presence of NaOH (aq) in ethanol to afford the corresponding 5-aryl-2,3-dihydrofuro[3,2-c]pyridin-4(5H)-ones in high yields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,3-Dihydrofuran, 99%