Merck
Molecular modelling of phthalates - PPARs interactions.
Journal of enzyme inhibition and medicinal chemistry (2008-09-30)
Nicolas Kambia, Nicolas Renault, Sebastien Dilly, Amaury Farce, Thierry Dine, Bernard Gressier, Michel Luyckx, Claude Brunet, Philippe Chavatte
ZUSAMMENFASSUNG
Di(2-ethylhexyl) phthalate (DEHP) is the most widely plasticizer for polyvinyl chloride (PVC) that is used in plastic tubes, in medical and paramedical devices as well as in food storage packaging. The toxicological profile of DEHP has been evaluated in a number of experimental animal models and has been extensively documented. Its toxicity is in part linked to the activation of the peroxisome proliferator-activated receptor alpha (PPAR(alpha)). As a response, an intensive research for a new, biologically inert plasticizer has been initiated. Among the alternative studied, tri(2-ethylhexyl) trimellitate (TEHTM) or trioctyl trimellitate (TOTM) has attracted increasing interest. However, very little information is available on their biological effects. We proceeded to dock TOTM, DEHP and its metabolites in order to identify compounds that are likely to interact with PPAR(alpha) and PPAR(gamma) binding sites. The results obtained hint that TOTM is not able to bind to PPARs and should therefore be safer than DEHP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trioctyltrimellitat, 99%